Alfa Aesar™ Diphenyl sulfoxide, 98+%

Manufacturer: Alfa Aesar™ B2213522

Quantity 100g

 View more versions of this product

Catalog No. AAB2213522

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 945-51-7
    Molecular Formula C12H10OS
    Formula Weight 202.28
    MDL Number MFCD00002085
    Beilstein 1908444
    Synonym Phenyl sulfoxide
    Chemical Name or Material Diphenyl sulfoxide

    Specifications

    Formula Weight 202.28
    TSCA TSCA
    MDL Number MFCD00002085
    Beilstein 1908444
    Synonym Phenyl sulfoxide
    Boiling Point 206° to 208°C (13mmHg)
    Melting Point 69° to 72°C
    Quantity 100g
    Molecular Formula C12H10OS
    CAS 945-51-7
    RTECS Number DA9185000
    Chemical Name or Material Diphenyl sulfoxide
    Assay ≥98%

    Description

    Applications
    Diphenyl sulfoxide is a reagent used as an oxidant and hydroxyl activator in combination with a variety of electrophilic reagents. Efficient glycosidation of a phenyl thiosialoside donor with diphenyl sulfoxide and triflic anhydride in dichloromethane was reported. Direct glycosylations with 1-hydroxy glycosyl donors is made using trifluoromethanesulfonic anhydride and diphenyl sulfoxide. The preparation of triarylsulfonium halides was by the action of aryl Grignard reagents on diphenyl sulfoxide.

    Solubility
    Soluble in water. Soluble in most organic solvents.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit