Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenylacetaldehyde, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
947-91-1
C14H12O
196.249
MFCD00006972
HLLGFGBLKOIZOM-UHFFFAOYSA-N
diphenylacetaldehyde,diphenylketen,acetaldehyde, diphenyl,alpha-phenylbenzeneacetaldehyde,diphenylethanal,diphenyl-acetaldehyde,benzeneacetaldehyde, .alpha.-phenyl,diphenyl-acetaldehyd,benzeneacetaldehyde, ?-phenyl,diphenylacetoaldehyde
13696
2,2-diphenylacetaldehyde
C1=CC=C(C=C1)C(C=O)C2=CC=CC=C2
947-91-1
C14H12O
196.249
MFCD00006972
HLLGFGBLKOIZOM-UHFFFAOYSA-N
diphenylacetaldehyde,diphenylketen,acetaldehyde, diphenyl,alpha-phenylbenzeneacetaldehyde,diphenylethanal,diphenyl-acetaldehyde,benzeneacetaldehyde, .alpha.-phenyl,diphenyl-acetaldehyd,benzeneacetaldehyde, ?-phenyl,diphenylacetoaldehyde
13696
2,2-diphenylacetaldehyde
C1=CC=C(C=C1)C(C=O)C2=CC=CC=C2
Density 1.1000g/mL
Packaging Glass bottle
Infrared Spectrum Authentic
Flash Point >110°C
Boiling Point 315.0°C
Quantity 5g
Refractive Index 1.5900 to 1.5920 (20°C, 589nm)
Specific Gravity 1.1
Color Colorless to Yellow
Assay Percent Range 97%
Linear Formula (C6H5)2CHCHO
Beilstein 07, 438
Formula Weight 196.25
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Diphenylacetaldehyde
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.