Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Diphenylacetylene-4,4'-diboronic acid bis(pinacol) ester, 95%

Manufacturer:  Alfa Aesar™ H5189703

Quantity 1g

 View more versions of this product

Catalog No. AAH5189703


  Check Availability
Add to cart

Specifications

Description

Applications
It is used as an active pharmaceutical intermediate.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Description & Specifications

Chemical Identifiers

849681-64-7
430.16
4,4'-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)diphenylacetylene
C26H32B2O4
MFCD16294524
Diphenylacetylene-4,4'-diboronic acid bis(pinacol) ester

Specifications

849681-64-7
430.16
4,4'-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)diphenylacetylene
Diphenylacetylene-4,4'-diboronic acid bis(pinacol) ester
C26H32B2O4
MFCD16294524
1g
95%
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit