Alfa Aesar™ Diphenylacetylene-4,4'-diboronic acid bis(pinacol) ester, 95%

Manufacturer: Alfa Aesar™ H5189706

Quantity 5g

 View more versions of this product

Catalog No. AAH5189706

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 849681-64-7
    Molecular Formula C26H32B2O4
    Formula Weight 430.16
    MDL Number MFCD16294524
    Synonym 4,4'-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)diphenylacetylene
    Chemical Name or Material Diphenylacetylene-4,4'-diboronic acid bis(pinacol) ester

    Specifications

    Formula Weight 430.16
    Molecular Formula C26H32B2O4
    MDL Number MFCD16294524
    CAS 849681-64-7
    Synonym 4,4'-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)diphenylacetylene
    Quantity 5g
    Chemical Name or Material Diphenylacetylene-4,4'-diboronic acid bis(pinacol) ester
    Assay 95%

    Description

    Applications
    It is used as an active pharmaceutical intermediate.

    Notes
    Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit