Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenylamine, 1% (w/v) in Sulfuric Acid, Ricca Chemical

Manufacturer:  Ricca Chemical CompanySupplier Diversity Partner 258416

Packaging glass sc amb
Quantity 473mL

 View more versions of this product

Catalog No. 258416

  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7664-93-9,122-39-4
100.0,0.55
Diphenylamine
97.0,0.53
Liquid

Specifications

7664-93-9,122-39-4
100.0,0.55
Liquid
Clear colorless
Diphenylamine
97.0,0.53
glass sc amb
1% (w/v) in Sulfuric Acid
473mL

Safety and Handling

ShelfLife : 12 months

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit