Learn More
Diphenylamine, ACS, Thermo Scientific Chemicals
Description
Diphenylamine is employed as harvest scald inhibitor for apples indoor drench treatments due to its antioxidant property. Alkyl derivatives of diphenylamine are used as anti-ozonants in the rubber manufacturing. It is used as lubricants and acts as a stabilizer for plastics. In the analytical chemistry, it is used to test the nitrate ion in association with ammonium chloride in sulfuric acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 52°C to 55°C |
Color | White |
Density | 1.16 g/mL |
Boiling Point | 302°C |
Flash Point | 152°C (305°F) |
Linear Formula | (C6H5)2NH |
Quantity | 100 g |
UN Number | UN3077 |
Beilstein | 508755 |
Sensitivity | Air and light sensitive |
Show More |
Safety and Handling
Hazard Category | H301+H311+H331-H373 |
Hazard Statement | GHS H Statement H301-H311-H331-H373 Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. May cause damage to organs through prolonged or repeated exposure. |
Precautionary Statement | P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P361-P363-P501c |
DOTInformation | Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S. |
EINECSNumber | 204-539-4 |
RTECSNumber | JJ7800000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.