Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenylamine, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
125g
500g
Packaging:
Wide Mouth Amber Glass Bottle
122-39-4
C12H11N
169.227
DMBHHRLKUKUOEG-UHFFFAOYSA-N
diphenylamine,n-phenylbenzenamine,anilinobenzene,benzenamine, n-phenyl,n,n-diphenylamine,phenylaniline,scaldip,n-phenylbenzeneamine,big dipper,no-scald
11487
CHEBI:4640
N-phenylaniline
C1=CC=C(C=C1)NC2=CC=CC=C2
122-39-4
C12H11N
169.227
DMBHHRLKUKUOEG-UHFFFAOYSA-N
diphenylamine,n-phenylbenzenamine,anilinobenzene,benzenamine, n-phenyl,n,n-diphenylamine,phenylaniline,scaldip,n-phenylbenzeneamine,big dipper,no-scald
11487
CHEBI:4640
N-phenylaniline
C1=CC=C(C=C1)NC2=CC=CC=C2

High quality chemicals for laboratory and specialized industrial use

Flash Point 150.00°C
Quantity 125g
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 1.16kg
Formula Weight 169.23
Physical Form Crystals
Grade BAKER ANALYZED™ ACS Reagent
Chemical Name or Material Diphenylamine