Alfa Aesar™ Diphenylborinic anhydride, 95%

Manufacturer: Alfa Aesar™ H5550906

Quantity 5g

 View more versions of this product

Catalog No. AAH5550906

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 4426-21-5
    Linear Formula (C6H5)2BOB(C6H5)2
    Formula Weight 346.04
    MDL Number MFCD00042663
    Beilstein 2950331
    Chemical Name or Material Diphenylborinic anhydride

    Specifications

    Formula Weight 346.04
    Melting Point 135° to 140°C
    Physical Form Powder
    MDL Number MFCD00042663
    CAS 4426-21-5
    Color White to yellow
    Sensitivity Moisture Sensitive
    Beilstein 2950331
    Quantity 5g
    Linear Formula (C6H5)2BOB(C6H5)2
    Chemical Name or Material Diphenylborinic anhydride
    Assay 95%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit