Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Diphenylcarbamyl chloride, 98%

Click to view available options
Quantity:
25g
100g
83-01-2
C13H10ClNO
231.679
MFCD00000633
XNBKKRFABABBPM-UHFFFAOYSA-N
diphenylcarbamyl chloride,diphenylcarbamoyl chloride,carbamic chloride, diphenyl,diphenylcarbamic chloride,n,n-diphenylcarbamyl chloride,diphenylchloroformamide,carbamoyl chloride, diphenyl,carabamic chloride, diphenyl,chloroformic acid diphenylamide,carbamic chloride, n,n-diphenyl
65741
N,N-diphenylcarbamoyl chloride
C1=CC=C(C=C1)N(C2=CC=CC=C2)C(=O)Cl
83-01-2
C13H10ClNO
231.679
MFCD00000633
XNBKKRFABABBPM-UHFFFAOYSA-N
diphenylcarbamyl chloride,diphenylcarbamoyl chloride,carbamic chloride, diphenyl,diphenylcarbamic chloride,n,n-diphenylcarbamyl chloride,diphenylchloroformamide,carbamoyl chloride, diphenyl,carabamic chloride, diphenyl,chloroformic acid diphenylamide,carbamic chloride, n,n-diphenyl
65741
N,N-diphenylcarbamoyl chloride
C1=CC=C(C=C1)N(C2=CC=CC=C2)C(=O)Cl
Applications
Diphenylcarbamoyl chloride was used to isolate peptido-keratan sulphate fragments from bovine intervertebral discs.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store away from oxidizing agents, water/ moisture, bases.
Melting Point 82°C to 86°C
Quantity 100g
Odor Irritating
UN Number UN3261
Beilstein 515312
Formula Weight 231.68
Percent Purity 98%
Chemical Name or Material Diphenylcarbamyl chloride

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

RTECSNumber : EY5065000

TSCA : TSCA