Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenylcarbazone TS, (U.S.P. Test Solution), Spectrum™

Click to view available options
Quantity:
100 mL
Packaging:
Amber Glass Bottle

D-138, 10329-15-4

Spectrum™ Diphenylcarbazone TS, (U.S.P. Test Solution) is a solution, sometimes designated with "TS or Test Solution", that is prepared according to the specifications of the USP Pharmacopeial Convention, for the purpose of conducting USP and National Formulary tests and assays.
CAS 538-62-5,64-17-5
CAS Max % 0.001,0.2
Molecular Formula C13H12N4O
UN Number UN1170
InChI Key ZFWAHZCOKGWUIT-UHFFFAOYSA-N
SMILES O=C(NNC1=CC=CC=C1)N=NC1=CC=CC=C1
IUPAC Name 3-(phenylamino)-1-(phenylimino)urea
Molecular Weight (g/mol) 240.27
Packaging Amber Glass Bottle
Quantity 100 mL