Diphenylcarbazone-Xylene Cyanol Mixed Indicator (not acidifier), for low level Chloride determination, Ricca Chemical

Manufacturer: Ricca Chemical CompanySupplier Diversity Partner 262016

Packaging Glass amber
Quantity 500mL

 View more versions of this product

Catalog No. 2620-16

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 64-17-5,538-62-5,2650-17-1,67-56-1 (Methanol (Methyl Alcohol))
    CAS Min % 98.0,0.31,0.04
    CAS Max % 100.0,0.32,0.04
    Physical Form Liquid
    Chemical Name or Material Diphenylcarbazone-Xylene Cyanol Mixed Indicator, Not acidifier

    Specifications

    Physical Form Liquid
    Color Clear blue-green
    Quantity 500mL
    Packaging Glass amber
    CAS 2650-17-1,67-56-1 (Methanol (Methyl Alcohol)),64-17-5,538-62-5
    CAS Max % 0.04,100.0,0.32
    CAS Min % 98.0,0.31,0.04
    Chemical Name or Material Diphenylcarbazone-Xylene Cyanol Mixed Indicator, Not acidifier

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit