Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Diphenyleneiodonium chloride, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
GPR3 agonist (EC50 = 1 μM); activates adenylate cyclase through GPR3 but not GPR6 or GPR12. Also induces Ca2+ mobilization and β-arrestin receptor internalization. Binds strongly to flavoproteins; inhibits several enzymes, including NO synthase, NADPH oxidases and NADPH cytochrome P450 oxidoreductase. Also inhibits platelet aggregation.
Specifications
Quantity | 10 mg |
Formula Weight | 314.55 |
Percent Purity | >99% |
Chemical Name or Material | Diphenyleneiodonium chloride |
Safety and Handling
Recommended Storage | Desiccate at -20°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction