Learn More
Diphenyliodonium chloride, 98+%, Thermo Scientific Chemicals
Description
Diphenyliodonium chloride is used as a reagent for electrophilic phenylation . It is used as a reactant for copper-catalyzed cross coupling reactions of purinesband arylation of anilines. It is involved in the preparation of acetylenic arylselenide and (arylmethylene)oxindoles. Further, it is employed in the preparation of aryl alkynes through Sonogashira coupling reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Diphenyliodonium chloride is used as a reagent for electrophilic phenylation . It is used as a reactant for copper-catalyzed cross coupling reactions of purinesband arylation of anilines. It is involved in the preparation of acetylenic arylselenide and (arylmethylene)oxindoles. Further, it is employed in the preparation of aryl alkynes through Sonogashira coupling reactions.
Solubility
Soluble in water and methanol.
Notes
Light sensitive. Incompatible with strong oxidizing agents.
Specifications
Melting Point | ∼230°C (decomposition) |
Quantity | 10 g |
UN Number | UN2811 |
Beilstein | 3574977 |
Sensitivity | Light sensitive |
Merck Index | 14,7464 |
Solubility Information | Soluble in water and methanol. |
Formula Weight | 316.57 |
Percent Purity | ≥98% |
Chemical Name or Material | Diphenyliodonium chloride |
Safety and Handling
Hazard Category | H301-H315-H319-H335-H500 |
Hazard Statement | GHS H Statement H301-H315-H319-H335 Toxic if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S. |
EINECSNumber | 216-049-8 |
RTECSNumber | NN6651666 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.