Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenyliodonium hexafluorophosphate, 97%, Acros Organics

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Plastic bottle
58109-40-3
C12H10I·F6P
426.081
DSSRLRJACJENEU-UHFFFAOYSA-N
diphenyliodonium hexafluorophosphate,diphenyliodanium hexafluorophosphate,diphenyliodonium hexafluorophosphate v,iodonium, diphenyl-, hexafluorophosphate 1-,iodonium, diphenyl-, hexafluorophosphate 1-1:1,diphenyliodonium pf6,dpihfp,acmc-209m2o,phi + pf6-,diphenyliodanium; hexafluoro-$l^ 5-phosphanuide
2737136
diphenyliodanium;hexafluorophosphate
C1=CC=C(C=C1)[I+]C2=CC=CC=C2.F[P-](F)(F)(F)(F)F
58109-40-3
C12H10I·F6P
426.081
DSSRLRJACJENEU-UHFFFAOYSA-N
diphenyliodonium hexafluorophosphate,diphenyliodanium hexafluorophosphate,diphenyliodonium hexafluorophosphate v,iodonium, diphenyl-, hexafluorophosphate 1-,iodonium, diphenyl-, hexafluorophosphate 1-1:1,diphenyliodonium pf6,dpihfp,acmc-209m2o,phi + pf6-,diphenyliodanium; hexafluoro-$l^ 5-phosphanuide
2737136
diphenyliodanium;hexafluorophosphate
C1=CC=C(C=C1)[I+]C2=CC=CC=C2.F[P-](F)(F)(F)(F)F
Melting Point 140.0°C to 144.0°C
Packaging Plastic bottle
Quantity 100g
Assay Percent Range 96% min. (HPLC)
Linear Formula (C6H5)2I+PF6-
Solubility Information Solubility in water: .. Other solubilities: soluble in methanol
Percent Purity 97%
Chemical Name or Material Diphenyliodonium hexafluorophosphate

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF SWALLOWED

GHS Signal Word: Danger

EINECSNumber : 261-134-5