Alfa Aesar™ Diphenyliodonium nitrate, 97%

Manufacturer: Alfa Aesar™ B2186106

Quantity 5g

 View more versions of this product

Catalog No. AAB2186106

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 722-56-5
    Molecular Formula C12H10INO3
    Formula Weight 343.12
    MDL Number MFCD00031704
    UN Number UN3087
    Beilstein 3922747
    Chemical Name or Material Diphenyliodonium nitrate

    Specifications

    Formula Weight 343.12
    Odor Odorless
    MDL Number MFCD00031704
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H272-H301-H315-H319-H335
    May intensify fire; oxidizer.
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Molecular Formula C12H10INO3
    CAS 722-56-5
    UN Number UN3087
    Beilstein 3922747
    Melting Point ≈150°C (dec.)
    Quantity 5g
    Health Hazard 3 GHS P Statement
    P221-P210-P301+P310-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: OXIDIZING SOLID, TOXIC, N.O.S.
    RTECS Number NN6665000
    Chemical Name or Material Diphenyliodonium nitrate
    Assay 97%

    GHS H Statement
    H272-H301-H315-H319-H335
    May intensify fire; oxidizer.
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P221-P210-P301+P310-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit