Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenylphosphinic Acid 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
1707-03-5
C12H11O2P
218.19
MFCD00002132
BEQVQKJCLJBTKZ-UHFFFAOYSA-N
phosphinic acid, diphenyl,hydroxydiphenylphosphine oxide,diphenylphosphinicacid,diphenyl phosphinic acid,diphenyl-phosphinic acid,acmc-1boli,di phenyl phosphinic acid,p,p-diphenylphosphinic acid,ksc180o2h,phosphinicacid, p,p-diphenyl
15567
CHEBI:37832
diphenylphosphinic acid
C1=CC=C(C=C1)P(=O)(C2=CC=CC=C2)O
1707-03-5
C12H11O2P
218.19
MFCD00002132
BEQVQKJCLJBTKZ-UHFFFAOYSA-N
phosphinic acid, diphenyl,hydroxydiphenylphosphine oxide,diphenylphosphinicacid,diphenyl phosphinic acid,diphenyl-phosphinic acid,acmc-1boli,di phenyl phosphinic acid,p,p-diphenylphosphinic acid,ksc180o2h,phosphinicacid, p,p-diphenyl
15567
CHEBI:37832
diphenylphosphinic acid
C1=CC=C(C=C1)P(=O)(C2=CC=CC=C2)O
Infrared Spectrum Authentic
Melting Point 193.0°C to 196.0°C
Color White
Quantity 5g
Packaging Glass bottle
Assay Percent Range 99%
Linear Formula (C6H5)2P(O)OH
Beilstein 16, 791
Formula Weight 218.19
Physical Form Fine Crystalline Solid
Percent Purity 99%
Chemical Name or Material Diphenylphosphinic acid, 99%
Show More Show Less

EINECSNumber : 216-948-5

RTECSNumber : SZ5315000