Learn More
Diphenylphosphinyl chloride, 98%, ACROS Organics™
Specifications
Infrared Spectrum | Authentic |
Color | Colorless to Yellow |
Density | 1.2400g/mL |
Quantity | 50g |
Flash Point | 26°C |
Specific Gravity | 1.24 |
Refractive Index | 1.6080 to 1.6120 |
Packaging | Glass bottle |
Boiling Point | 222.0°C (16.0 mmHg) |
Assay Percent Range | 97.5% min. (GC) |
Show More |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 216-107-2