Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Diphenylsilanediol, 98%

Manufacturer:  Alfa Aesar™ A1049214

Quantity 25g

 View more versions of this product

Catalog No. AAA1049214

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Description

Applications
Diphenylsilanediol is used in Medicine field, electronics industry and polymer material.

Solubility
Reacts with water.

Notes
Stable under normal temperatures and pressures. Incompatible with Oxidizing agents. Store at room temperature. Air & Light Sensitive.
Description & Specifications

Chemical Identifiers

947-42-2
(C6H5Si(OH)2
MFCD00002101
2523445
Diphenylsilanediol
C12H12O2Si
216.31
UN1325
Dihydroxydiphenylsilane

Specifications

947-42-2
(C6H5Si(OH)2
MFCD00002101
2523445
Odorless
25g
98%
C12H12O2Si
216.31
UN1325
Dihydroxydiphenylsilane
≈150°C (dec.)
Diphenylsilanediol

Safety and Handling

SDS & Safety Documents

GHS H Statement
H228
Flammable solid.

GHS P Statement
P210
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.

Warning

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.

RTECSNumber : VV3640000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit