Alfa Aesar™ Diphenyltin dichloride, 95%

Manufacturer: Alfa Aesar™ 7113904

Quantity 2g

 View more versions of this product

Catalog No. AA7113904

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1135-99-5
    Molecular Formula C12H10Cl2Sn
    Linear Formula (C6H5)2SnCl2
    Formula Weight 343.81
    MDL Number MFCD00000516
    Synonym Dichlorodiphenyltin
    Chemical Name or Material Diphenyltin dichloride

    Specifications

    Formula Weight 343.81
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Boiling Point 333° to 337°C (dec.)
    Linear Formula (C6H5)2SnCl2
    Flash Point >150°C (302°F)
    Color White
    Melting Point 41° to 43°C
    CAS 1135-99-5
    Health Hazard 2 GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    RTECS Number WH7253000
    Sensitivity Moisture Sensitive
    Physical Form Crystalline
    Molecular Formula C12H10Cl2Sn
    MDL Number MFCD00000516
    Quantity 2g
    Synonym Dichlorodiphenyltin
    Health Hazard 1 Warning
    Chemical Name or Material Diphenyltin dichloride
    Assay 95%

    GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit