Alfa Aesar™ Diphenyltin oxide

Manufacturer: Alfa Aesar™ 3961503

Quantity 1g

 View more versions of this product

Catalog No. AA3961503

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2273-51-0
    Formula Weight 288.9
    MDL Number MFCD00058851
    UN Number UN3146
    Synonym Oxodiphenyltin
    Chemical Name or Material Diphenyltin oxide

    Specifications

    Formula Weight 288.9
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Quantity 1g
    Melting Point 320°C (dec.)
    UN Number UN3146
    Synonym Oxodiphenyltin
    Health Hazard 1 Danger
    TSCA TSCA
    Physical Form Powder
    MDL Number MFCD00058851
    CAS 2273-51-0
    Health Hazard 2 GHS H Statement
    H331-H302-H312-H315-H319-H335
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.
    Chemical Name or Material Diphenyltin oxide

    GHS H Statement
    H331-H302-H312-H315-H319-H335
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit