Learn More
Diphtheria Toxin A Mouse anti-Bacteria, DyLight 680, Clone: 8A4, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NB1102565FR
Catalog No. NB1102565FR
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Diphtheria Toxin A Monoclonal antibody specifically detects Antigen in Bacteria samples. It is validated for Enzyme Linked Immunosorbent Assay.
Specifications
Specifications
Diphtheria Toxin A
8A4
ELISA, Sandwich ELISA
Purified
Mouse
IgG2a
0.1 ml
Store at 4C in the dark.
Monoclonal
Bacteria
ELISA
DyLight 680
50mM Sodium Borate with 0.05% Sodium Azide
Diphtheria Toxin
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Diphtheria toxoid.
Protein A purified
RUO
Primary
This reacts with free A subunits of Diphtheria toxin. It does not react with the whole Diphtheria toxin or free B subunit of Diphtheria toxin.