Learn More
Diphtheria Toxoid Mouse anti-Bacteria, Alexa Fluor™ 532, Clone: 17.6B9, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP223596AF532
Catalog No. NP22396A32
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Diphtheria Toxoid Monoclonal antibody specifically detects Diphtheria Toxoid in Bacteria samples. It is validated for ELISA.
Specifications
Specifications
Diphtheria Toxoid
17.6B9
ELISA
C. Diphtheria toxin, Corynebacterium diphtheriae toxin, Corynebacterium diphtheriae toxoid, DIP0222, Diptheria toxin, Diptheriae toxin, DTx
Diphtheria toxoid prepared from toxin purified from C. diphtheriae subsp. intermedius Park-Williams no. 8 by formaldehyde inactivation (2), and adsorbed to aluminum hydroxide gel.
0.1 mL
Primary
It reacts with diphtheria toxoid and also neutralizes diphtheria toxin as determined in a VERO cell assay
ELISA
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
Bacteria