Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DIPPA hydrochloride, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg

Chemical Identifiers

155512-52-0
C22H24Cl3N3OS
484.864
BNWYENYHNOESCX-ZMBIFBSDSA-N
2-(3, 4-Dichlorophenyl)-N-methyl-N-[(1S)-1-(3-isothiocyanatophenyl)-2-(1-pyrrolidinyl)ethyl]acetamide hydrochloride
45073425

Selective irreversible κ antagonist

An irreversible and selective antagonist at the κ receptor, with persistent effect in vivo.
Quantity 50mg
SMILES CN(C(CN1CCCC1)C2=CC(=CC=C2)N=C=S)C(=O)CC3=CC(=C(C=C3)Cl)Cl.Cl
IUPAC Name 2-(3,4-dichlorophenyl)-N-[(1S)-1-(3-isothiocyanatophenyl)-2-pyrrolidin-1-ylethyl]-N-methylacetamide;hydrochloride
Formula Weight 484.87
Chemical Name or Material DIPPA hydrochloride
Grade >99%

Recommended Storage : Desiccate at -20°C