Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dipropylamine, 99%, ACROS Organics™

Click to view available options
Quantity:
250mL
1L
Packaging:
Glass bottle
142-84-7
C6H15N
101.19
MFCD00009362
WEHWNAOGRSTTBQ-UHFFFAOYSA-N
dipropylamine,di-n-propylamine,1-propanamine, n-propyl,n-dipropylamine,n-propyl-1-propanamine,rcra waste number u110,dipropyl amine,rcra waste no. u110,unii-60p318iiry,ccris 4805
8902
N-propylpropan-1-amine
CCCNCCC
142-84-7
C6H15N
101.19
MFCD00009362
WEHWNAOGRSTTBQ-UHFFFAOYSA-N
dipropylamine,di-n-propylamine,1-propanamine, n-propyl,n-dipropylamine,n-propyl-1-propanamine,rcra waste number u110,dipropyl amine,rcra waste no. u110,unii-60p318iiry,ccris 4805
8902
N-propylpropan-1-amine
CCCNCCC
Viscosity 0.5 mPa.s (20°C)
Boiling Point 105.0°C to 110.0°C
Melting Point -63.0°C
Color Undesignated
Density 0.7400g/mL
Water 0.2% max.
Specific Gravity 0.74
Packaging Glass bottle
Flash Point 7°C
Refractive Index 1.4030 to 1.4050
Quantity 250mL
Infrared Spectrum Authentic
Assay Percent Range 99%
Linear Formula (CH3CH2CH2)2NH
Beilstein 04, 138
Merck Index 15, 3383
Solubility Information Solubility in water: 46g/L (20°C). Other solubilities: freely soluble in alcohol
Formula Weight 101.19
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material Dipropylamine, 99%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
May cause respiratory irritation.
Highly flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 205-565-9

RTECSNumber : JL9200000

TSCA : TSCA