Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dipyridamole, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
1 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
58-32-2
C24H40N8O4
504.64
IZEKFCXSFNUWAM-UHFFFAOYSA-N
2-({6-[bis(2-hydroxyethyl)amino]-4,8-bis(piperidin-1-yl)pyrimido[5,4-d][1,3]diazin-2-yl}(2-hydroxyethyl)amino)ethan-1-ol
OCCN(CCO)C1=NC(N2CCCCC2)=C2N=C(N=C(N3CCCCC3)C2=N1)N(CCO)CCO
58-32-2
C24H40N8O4
504.64
IZEKFCXSFNUWAM-UHFFFAOYSA-N
2-({6-[bis(2-hydroxyethyl)amino]-4,8-bis(piperidin-1-yl)pyrimido[5,4-d][1,3]diazin-2-yl}(2-hydroxyethyl)amino)ethan-1-ol
OCCN(CCO)C1=NC(N2CCCCC2)=C2N=C(N=C(N3CCCCC3)C2=N1)N(CCO)CCO

D1250, 58-32-2, C24H40N8O4

Spectrum™ Dipyridamole, USP is an antiplatelet drug. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Quantity 1 kg
Ignition Residue 0.001
Loss on Drying 0.002
CAS Max % 1
Percent Purity 98 to 102%