Learn More
Aviva Systems Biology Corp DIRAS1 Antibody - middle region (ARP52803_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP52803_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DIRAS1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human DIRAS1. Alternate names: Di-Ras1, FLJ42681, GBTS1, RIGApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rat, Zebrafish
Catalog No. 50-014-60837