Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DIRAS1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA550652
Description
The antibody detects endogenous levels of total DIRAS1 protein.
DIRAS1 belongs to a distinct branch of the functionally diverse Ras superfamily of monomeric GTPases.
Specifications
DIRAS1 | |
Polyclonal | |
Unconjugated | |
Diras1 | |
DIRAS family GTP binding RAS like 1; DIRAS family GTPase 1; DIRAS family, GTP-binding RAS-like 1; DIRAS1; Di-Ras1; Distinct subgroup of the Ras family member 1; GBTS1; GTP-binding protein Di-Ras1; ras-related inhibitor of cell growth; RIG; small GTP-binding tumor suppressor 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
148252 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
2.4 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
O95057 | |
Diras1 | |
Synthetic peptide corresponding to residues near the C terminal of human DIRAS family, GTP-binding RAS-like 1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction