Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DIRAS3 Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP282085AF700
This item is not returnable.
View return policy
Description
DIRAS3 Polyclonal antibody specifically detects DIRAS3 in Human samples. It is validated for Western Blot, ELISASpecifications
DIRAS3 | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, ELISA | |
Alexa Fluor 700 | |
DIRAS family, GTP-binding RAS-like 3, Distinct subgroup of the Ras family member 3, NOEY2member I, RHOI, Rho-related GTP-binding protein RhoI | |
DIRAS3 antibody was raised against an 18 amino acid peptide near the carboxy terminus of human DIRAS3. The immunogen is located within the last 50 amino acids of DIRAS3. Amino Acid Squence: TTGLQEPEKKSQMPNTTE | |
0.1 mL | |
Cell Cycle and Replication | |
9077 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction