Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DIRC1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA567811
Description
DIRC1 Polyclonal Antibody for Western Blot
This gene consists of two exons and encodes a 104aa protein. Disruption of this gene by translocation t(2;3)(q33;q21) is associated with familial clear cell renal cancer.
Specifications
DIRC1 | |
Polyclonal | |
Unconjugated | |
DIRC1 | |
DIRC1; disrupted in renal cancer protein; disrupted in renal carcinoma 1; disrupted in renal carcinoma protein 1 | |
Rabbit | |
affinity chromatography | |
RUO | |
116093 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q969H9 | |
DIRC1 | |
Synthesized peptide derived from human DIRC1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction