Learn More
Aviva Systems Biology Corp DIRC2 Antibody - middle region (ARP50133_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP50133_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DIRC2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human DIRC2. Alternate names: FLJ14784, RCC4ApplicationWB. Species reactivity: Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60264