Learn More
Sklar InstrumentsSupplier Diversity Partner Director and Tongue Tie, Sklar, 8 in., Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 301080
Director and Tongue Tie, Sklar, 8 in., Stainless Steel. Sklar's Director and Tongue Tie is commonly used in examining the mouth of a pediatric patient. It is also used in the treatment of ankyloglossia, or in.tongue tie. in. One end of the instrument is flat, and the other can be pointed or have a small, round probe. The flat end is the tongue tie. It is pear shaped with a central split from the distal end. The split in the flat end assists in isolating and stabilizing the tongue or the lingual frenulum during procedures.
Catalog No. 50-143-8096