Alfa Aesar™ Dirhodium(II) tetrakis(caprolactam), complex with acetonitrile (1:2)

Manufacturer: Alfa Aesar™ 44686MB

Quantity 25mg

 View more versions of this product

Catalog No. AA44686MB

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 138984-26-6
    Molecular Formula C24H40N4O4Rh2·2CH3CN
    Formula Weight 736.52
    MDL Number MFCD00467690
    Synonym Rh2(capy)4
    Chemical Name or Material Dirhodium (II) tetrakis (caprolactam), Complex with acetonitrile (1:2)

    Specifications

    Formula Weight 736.52
    Synonym Rh2(capy)4
    Physical Form Powder
    Molecular Formula C24H40N4O4Rh2·2CH3CN
    MDL Number MFCD00467690
    CAS 138984-26-6
    Color Purple
    Quantity 25mg
    Chemical Name or Material Dirhodium (II) tetrakis (caprolactam), Complex with acetonitrile (1:2)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit