Alfa Aesar™ Dirhodium(II) tetrakis(methyl 2-pyrrolidinone-5(S)-carboxylate)acetonitrile, 2-propanol complex, Rh nominally 25.5%

Manufacturer: Alfa Aesar™ 4463802

Quantity 0.5g

 View more versions of this product

Catalog No. AA4463802

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 132435-65-5
    Molecular Formula C24H32N4O12Rh2·2CH3CN·CH3CH(OH)CH3
    Formula Weight 871.46
    MDL Number MFCD00192123
    Synonym Rh2(5S-MEPY)4
    Chemical Name or Material Dirhodium (II) tetrakis (methyl 2-pyrrolidinone-5 (S)-carboxylate)acetonitrile, 2-propanol complex

    Specifications

    Formula Weight 871.46
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Powder
    Molecular Formula C24H32N4O12Rh2·2CH3CN·CH3CH(OH)CH3
    CAS 132435-65-5
    MDL Number MFCD00192123
    Synonym Rh2(5S-MEPY)4
    Health Hazard 1 Danger
    Concentration Rh nominally 25.5%
    Health Hazard 2 GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Storage Note 1
    • Packaged under Argon
    Quantity 0.5g
    Chemical Name or Material Dirhodium (II) tetrakis (methyl 2-pyrrolidinone-5 (S)-carboxylate)acetonitrile, 2-propanol complex

    GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    • Packaged under Argon

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit