Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DIS3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA578427
Description
Positive Control: U87-MG, SK-N-SH, IMR-32, SK-N-AS Predicted reactivity: Human (98%), Mouse (89%), Rat (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The exosome is involved in a multitude of cellular RNA processing and degradation events. DIS3, also known as exosome complex exonuclease RRP44, is a ribonuclease that acts directly in the processing, turnover, and surveillance of a large number of distinct RNA species. DIS3 localizes to both the cytoplasm and the nucleus and contains one PINc domain. It is widely expressed with highest expression in testis and is required for processing of 7S pre-RNA into a mature nuclear complex and, ultimately, for proper mitotic progression. Abnormal expression levels of DIS3 may be associated with colon cancer, suggesting a role for DIS3 in tumorigenesis.
Specifications
DIS3 | |
Polyclonal | |
Unconjugated | |
DIS3 | |
2810028N01Rik; bA555G22.1; DIS3; DIS3 exosome endoribonuclease and 3'-5' exoribonuclease; DIS3 homolog, exosome endoribonuclease and 3'-5' exoribonuclease; DIS3 mitotic control homolog; DIS3 mitotic control homolog (S. cerevisiae); dis3p; EXOSC11; exosome complex exonuclease RRP44; exosome component 11; KIAA1008; mitotic control protein dis3 homolog; protein DIS3 homolog; RGD1304646; Ribosomal RNA-processing protein 44; RP11-342J4.3; RRP44 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22894, 306103, 72662 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9CSH3, Q9Y2L1 | |
DIS3 | |
Recombinant protein encompassing a sequence within the N-terminus region of human DIS3. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction