Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DIS3L Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598648
Description
DIS3L Polyclonal Antibody for Western Blot, IHC (P), ELISA
DIS3L is a probable exonuclease. It has four isoforms.
Specifications
DIS3L | |
Polyclonal | |
Unconjugated | |
DIS3L | |
AV340375; DIS3 like exosome 3'-5' exoribonuclease; DIS3 mitotic control homolog (S. cerevisiae)-like; DIS3 mitotic control homolog-like; Dis3l; DIS3L1; DIS3-like exonuclease 1; DIS3-like exosome 3'-5' exoribonuclease; KIAA1955; RGD1308959 | |
Rabbit | |
Protein G | |
RUO | |
115752 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
3 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q8TF46 | |
DIS3L | |
Recombinant Human DIS3-like exonuclease 1 protein (1-300AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction