Learn More
DIS3L2 Mouse anti-Human, DyLight 680, Clone: 6C7B2, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237316FR
Catalog No. NBP237316FR
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
DIS3L2
6C7B2
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry
Purified
DIS3L2
Purified recombinant fragment of human DIS3L2 (AA: 27-250) expressed in E. Coli.
Protein A or G purified
Store at 4C in the dark.
Monoclonal
Human
ELISA, Flow Cytometry, Immunohistochemistry, Western Blot
DyLight 680
50mM Sodium Borate with 0.05% Sodium Azide
DIS3 mitotic control homolog (S. cerevisiae)-like 2, DIS3-like exonuclease 2, FAM6A, member A
Mouse
IgG1
0.1 ml
Primary
129563