Learn More
Dynalon Discs for 2 piece Plastic Buchner Funnels
Manufacturer: Dynalon 242845070
Description
Made from high-density polyethylene, these discs have excellent chemical resistance. They are useful for holding up filter paper. In this way, the blocking of the holes is avoided and the filtering surface of the funnel is free. Will withstand temperatures up to 110°C.
Order Info
Supplied in packages of 10 discs. 50 discs per case. Mesh size 950 x 700 microns.
Specifications
Filter Paper | |
HDPE | |
1.75in | |
1.76in | |
Buchner Funnel |
10 per pack | |
Disc | |
7.01cm | |
70mm |