Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dishevelled-1 Mouse anti-Human, Mouse, Rat, Clone: 1-3F12, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP261609

Catalog No. NBP261609

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

Dishevelled-1
1-3F12
Unconjugated
PBS with 0.02% Sodium Azide
dishevelled 1 (homologous to Drosophila dsh), dishevelled, dsh homolog 1 (Drosophila), Dishevelled-1, DSH homolog 1, DVL, DVL1L1, MGC54245, segment polarity protein dishevelled homolog DVL-1
Mouse
Protein A purified
Signal Transduction, Wnt Signaling Pathway
Primary
1855
Immunoprecipitation, Western Blot
1 mg/mL
Western Blot 1:200, Immunoprecipitation 1:10-1:500
Purified
DVL1
GST fusion protein containing the 45 carboxy-terminal amino acids of Dishevelled-1.
0.1 ml
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit