Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dishevelled-2 Mouse anti-Human, Alexa Fluor™ 700, Clone: OTI2D11, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270611AF700

Catalog No. N270611A700


Add to cart

Description

Description

Dishevelled-2 Monoclonal antibody specifically detects Dishevelled-2 in Human samples. It is validated for Immunohistochemistry.
Specifications

Specifications

Dishevelled-2
OTI2D11
Immunohistochemistry
Affinity Purified, Conjugated
DVL2
Recombinant protein fragment corresponding to amino acids 1-320 of human Dishevelled-2 (NP_004413) produced in E.coli.
Immunogen affinity purified
Phospho Specific, Signal Transduction, Wnt Signaling Pathway
Primary
1856.0
Immunohistochemistry
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
dishevelled 2 (homologous to Drosophila dsh), dishevelled, dsh homolog 2 (Drosophila), Dishevelled-2, DSH homolog 2, segment polarity protein dishevelled homolog DVL-2
Mouse
IgG2b
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit