Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Disodium 8-Amino-1,3,6-naphthalenetrisulfonate Hydrate 99.0+%, TCI America™

Click to view available options
Quantity:
25g
500g
5398-34-5
C10H9NNa2O9S3
429.34
MFCD00070572
CLVNIGIUQSLKER-UHFFFAOYSA-N
Amino H Acid Disodium Salt, 8-Amino-1,3,6-naphthalenetrisulfonic Acid Disodium Salt, Koch Acid Disodium Salt, 1-Naphthylamine-3,6,8-trisulfonic Acid Disodium Salt
71431007
8-aminonaphthalene-1,3,6-trisulfonic acid;sodium
C1=C2C=C(C=C(C2=C(C=C1S(=O)(=O)O)N)S(=O)(=O)O)S(=O)(=O)O.[Na].[Na]
5398-34-5
C10H9NNa2O9S3
429.34
MFCD00070572
CLVNIGIUQSLKER-UHFFFAOYSA-N
Amino H Acid Disodium Salt, 8-Amino-1,3,6-naphthalenetrisulfonic Acid Disodium Salt, Koch Acid Disodium Salt, 1-Naphthylamine-3,6,8-trisulfonic Acid Disodium Salt
71431007
8-aminonaphthalene-1,3,6-trisulfonic acid;sodium
C1=C2C=C(C=C(C2=C(C=C1S(=O)(=O)O)N)S(=O)(=O)O)S(=O)(=O)O.[Na].[Na]
Color White-Yellow
Quantity 500g
Formula Weight 427.32
Physical Form Crystal-Powder at 20°C
Percent Purity ≥99.0% (HPLC,T)
Chemical Name or Material Disodium 8-Amino-1,3,6-naphthalenetrisulfonate Hydrate