Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Disodium Dihydrogen Ethylenediaminetetraacetate Dihydrate 99.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
6381-92-6
C10H18N2Na2O10
372.238
MFCD00150037
OVBJJZOQPCKUOR-UHFFFAOYSA-L
edta disodium salt,cal-ex decalcifier,buffer solution, ph 10.00,sodium di ethylenediamine tetraacetate dihydrate,ethylenediamine tetraacetic acid, disodium salt dihydrate,ethylenediamine tetraacetic acid, disodium salt, standard solution,sodium di ethylenediamine tetraacetate standard solution,ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs
44120005
disodium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;dihydrate
C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)O)CC(=O)O.O.O.[Na+].[Na+]
6381-92-6
C10H18N2Na2O10
372.238
MFCD00150037
OVBJJZOQPCKUOR-UHFFFAOYSA-L
edta disodium salt,cal-ex decalcifier,buffer solution, ph 10.00,sodium di ethylenediamine tetraacetate dihydrate,ethylenediamine tetraacetic acid, disodium salt dihydrate,ethylenediamine tetraacetic acid, disodium salt, standard solution,sodium di ethylenediamine tetraacetate standard solution,ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs
44120005
disodium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;dihydrate
C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)O)CC(=O)O.O.O.[Na+].[Na+]
Quantity 25g
Melting Point 252°C
Color White
Formula Weight 336.21
Physical Form Crystal-Powder at 20°C
Percent Purity ≥99.0% (T)
Chemical Name or Material Disodium Dihydrogen Ethylenediaminetetraacetate Dihydrate [for Biochemical Research]

EINECSNumber : (2)-1265

RTECSNumber : AH4410000

TSCA : Yes