Learn More
Disorbent, Gas Analysis Chemcials, Burrell Scientific
Manufacturer:  Burrell Scientific Inc 0397300016
Catalog No. 14-400-153
Add to cart
Description
Description
  • Used for the determination of CO2  values in a test sample
  • Also absorbs other acid gases such as H2S, HCl, SO2
Recommended Storage
Store at room temperature
Specifications
Specifications
Disorbent, Gas Analysis Chemcials
50-70%, 30-50%
Caustic potash-liquid or solutions
Water: Completely Soluble
2mm Hg 45% Solution at 20°C (68°F)
Colorless
13.5 (0.1M soln.)
1310-58-3, 7732-18-5
(10% Solution: 101°C (213°F)), (45% Solution: 132°C (270°F)
Liquid
(10% Solution: 1.1), (45% Solution: 1.45)
340°C (644°F)
-3°C (10% Solution), -29°C (45% Solution)
16 oz.