Learn More
Abnova™ Dispase (B. polymyxa)
Used for Enzyme
Manufacturer:  Abnova™ P5317
Catalog No. 89-942-015
Add to cart
Description
Description
#REF!
Specifications
Specifications
P29148
Lyophilized
Dispase (B. polymyxa)
10 mg
RUO
Neutral Protease (Dispase) Assay; Tissue disaggregation and subcultivation; Prevention of unwanted cell clumping; Preparation of cells for culture; Separation of intact epidermis from dermis and intact epithelial sheet in culture from the substratum; Harvest and transfer of normal, diploid cells and cell lines; Gentle and intact detachment of epidermal cells.
≥ 4 units per mg dry weight. One Unit releases Folin positive amino acids equivalent to 1μmole tyrosine per minute from casein at 37°C, pH 7.5.
None
Enzyme Activity
36kDa
Native protein chromatographically purified from Bacillus polymyxa
Store at 4°C.
DISPASE
Native protein chromatographically purified from Bacillus polymyxa
Bacteria