BrandTech™ Dispensette™ S Analog-adjustable Bottletop Dispensers

The Dispensette™ S Analog Bottletop Dispensers feature a “floating piston” design for low-force operation and no wearing parts. Optimized flow channels make dispensing and priming even easier-particularly in instruments with large volumes. Designed for general-purpose dispensing applications.

Manufacturer: BrandTech™ 4600161

 View more versions of this product

Catalog No. 13-689-017

  Check Availability
Add to Cart

    Specifications

    Volume Range 5-50mL
    Color Red
    Display Analog
    Product Type Dispenser
    Increments 1.0mL
    Autoclavable At 121°C
    Accuracy 0.5% (250μL)
    Description With recirculation valve
    Item Description Applications: Dispensing aggressive reagents, including concentrated bases and acids such as H3PO4, H2SO4 (except e.g., HCl, HNO3 und HF), saline solutions, as well as many organic solvents
    Certifications/Compliance ISO 9001

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit