Learn More
BrandTech™ BRAND™ Dispensing Cylinder with Valve Block
For use with Titrette™ Bottletop Burette. Accommodates all serial number ranges.
$225.00
Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
13-690-104
|
BrandTech™ BRAND™
707533 |
10 mL Titrette bottletop burette |
Each of 1 for $225.00
|
|
Description
- For use with Titrette™ Bottletop Burette
- Fits all serial number ranges
- *For 50mL instruments with serial numbers beginning with 01-12 AND letters A-J, must order both Dispensing cylinder and Titrating tube as a pair.
Specifications
Dispensing Cylinder | |
Valve head for titrette |