Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Disperse Red 19, pure, Acros Organics

Click to view available options
Quantity:
50g
2734-52-3
C16H18N4O4
330.344
MFCD00044367
GHDZRIQTRDZCMV-UHFFFAOYSA-N
C.I. 11130
17651
2-[N-(2-hydroxyethyl)-4-[(4-nitrophenyl)diazenyl]anilino]ethanol
C1=CC(=CC=C1N=NC2=CC=C(C=C2)[N+](=O)[O-])N(CCO)CCO
2734-52-3
C16H18N4O4
330.344
MFCD00044367
GHDZRIQTRDZCMV-UHFFFAOYSA-N
C.I. 11130
17651
2-[N-(2-hydroxyethyl)-4-[(4-nitrophenyl)diazenyl]anilino]ethanol
C1=CC(=CC=C1N=NC2=CC=C(C=C2)[N+](=O)[O-])N(CCO)CCO
Dye Content 20% min.
Infrared Spectrum Authentic
Formula Weight 330.33
Physical Form Crystalline Powder
Grade Extra Pure
UV Spectrometry Complies
Color Brown-Red to Purple
Quantity 50g
Chemical Name or Material Disperse red 19