Learn More
Ted Pella Inc Disposable Base Molds, 15 x 15 x 5mm, 500/box
Supplier: Ted Pella Inc 271472
Disposable base molds offer ease and convenience of operation. They are inexpensive enough to be discarded after use, yet strong enough to be reused. Made of PVC, they offer excellent thermal exchange. They have a smooth interior finish and rounded corners to facilitate their removal. Also, they are available in the same variety of sizes as metal molds and can be used with the same styles or types of cassettes and embedding rings.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.