Thermo Scientific™ Disposable Blade Carriers
Utilize low- and high-profile blades with the quick clamp system of Thermo Scientific™ Disposable Blade Carriers.
Manufacturer: Thermo Scientific™ 705100
 View more versions of this product
Catalog No. 70-510-0
  Check Availability
Add to Cart
    Specifications
    Type Disposable Blade Carrier ER
    Description ER Type blade carrier for dovetail style units
    Item Description Disposable blade carrier ER
    For Use With (Equipment) Dovetail Style Units
    Utilize low- and high-profile blades with the quick clamp system of Thermo Scientific™ Disposable Blade Carriers. Updated models (long stroke and version 2 bar guideways) include removable angle adjustment and blade clamp levers.
    Disposable Blade Carriers
    • Blade angle and blade adjustment levers are removable to help protect user settings (Version 2 and 3 units only)
    • Concentric lever locks onto the dovetail to help reduce accidental movement of the holder (Version 2 and 3 units only)
    • Quick clamp system for high- and low-profile blades
    • Adjustable clearance angle
    • Moveable knife guard covers the blade entirely in any position
    • Knife carrier base included
  • Description & Specifications
Related Products
Product Content Feedback