Learn More
United Scientific™ Disposable Borosilicate Glass 7.0 Pasteur Pipets
SDPSCI_ED
Borosilicate Glass Pasteur Pipet offers an affordable alternative.
$122.00
Specifications
Material Borosilicate Glass
Sterility Non-sterile
Graduated No
Color Clear
Type Pasteur Pipet
View More Specs
Products 2
Catalog Number Mfr. No. Length (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Length (Metric) Price Quantity & Availability  
S00357
SDPSCI_ED
View Documents
United Scientific™
DBG146NS
146 mm
Case of 1000 for $122.00
Add to Cart
S00358
SDPSCI_ED
View Documents
United Scientific™
DBG229NS
229 mm
Case of 1000 for $122.00
Add to Cart
Description
Description
United Scientific Pasteur pipettes are manufactured to conform strict specifications laid down by research and industrial laboratories and are suitable for various laboratory applications.
  • Available in 2 lengths
  • Made from Borosilicate Glass
  • Facilitate Controlled Drop
  • Smooth, Straight-Cut, Fire-Polished Tips
  • Ideal for Clinical and Industrial Use
  • Disposable
Specifications
Specifications
Borosilicate Glass
No
Pasteur Pipet
Non-sterile
Clear
Single-use