Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ Disposable Frocks
White 58 gsm polypropylene and polyethylene frocks that create a barrier against particles, micro-organisms and nonhazardous liquids.
Description
- Front zipper closure
- Elastic cuffs
- Snap-closure collar
- Knee-length coverage
Specifications
Knee-length Frock | |
Polypropylene, Polyethylene | |
Zipper | |
Yes | |
58 gsm | |
115 cm | |
65 cm |
X-Large | |
White | |
Elastic | |
Non-sterile | |
45 in. | |
26 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title